Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Protein S Containing Glycine and NaCl

Catalog No. ICN19408180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19408180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19408180 Supplier MP Biomedicals™ Supplier No. 0219408180
Only null left
Add to Cart
Add to Cart

From human plasma

A vitamin K-dependent protein which is a cofactor of activated protein C (APC). It increases the affinity of APC for phospholipids which leads to factor V and VIII inhibition. Plays an essential role in the regulation of the coagulation system.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Regulation of the Coagulation System
Molecular Weight (g/mol) 69000.5
Name Protein S
Preparation Method Prepared from Human Plasma that has been shown to be negative for HBsAg and for Antibodies to HIV and HCV
Quantity 100 μg
Source Human Plasma
Storage Requirements 0°C
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.