Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ratchet Bolt for FastPrep-24

Catalog No. MP116002511 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep 24 all models
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
MP116002511 FastPrep 24 all models
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116002511 Supplier MP Biomedicals™ Supplier No. 116002511
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Securing screw for FastPrep-24™

FastPrep accessory for securing various adapters to FastPrep-24 instruments.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) FastPrep 24 all models
Packaging Quantity 1 Ea.
Product Type Ratchet Bolt
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.