Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ready to Use 5% 7X™ Solutions

Catalog No. ICN7668493 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Description:
7X-O-Matic; 1 gal.
7X; 1 gal.
ES 7X; 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
ICN7668493 7X-O-Matic; 1 gal.
ICN7668093 7X; 1 gal.
ICN7668193 ES 7X; 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN7668493 Supplier MP Biomedicals™ Supplier No. 097668493
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 1 gallon Ready to Use 5% solution of 7X that includes a hand-pump for convenient dispensing

  • Ideal for glass and plastic labware
  • Contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware
  • Environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Concentration 7X
Content And Storage 15°C to 30°C
Description 7X-O-Matic; 1 gal.
For Use With (Application) For Glass, Plastic, benchtop equipment and tubing
Volume (English) 1 gal.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.