Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RNaid™ Kit

Catalog No. MP111107200 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Product Type:
Slurry
Slurry plus SPIN Filter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
MP111107200 Slurry plus SPIN Filter
MP111007200 Slurry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP111107200 Supplier MP Biomedicals™ Supplier No. 111107200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RNaid™ Kit provides a simple and convenient way to isolate total RNA from agarose and polyacrylamide gels and from solutions.

  • Kit is designed to isolate and purify RNA (75 bases to 10 kb) from native and denaturing agarose and polyacrylamide gels.
  • Removal of unincorporated nucleotides, buffer salts, and other reagents from enzyme reactions.
  • The purified RNA is ready for reverse transcription and PCR, labeling and microarray analysis, and other common applications.
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Enzymatic Reactions, Solutions, Agarose Gels, Polyacrylamide Gels
For Use With (Application) To isolate, purify RNA (75 bases to 10 kb) from native, denaturing agarose/polyacrylamide gels
Product Type Slurry plus SPIN Filter
Quantity 200 Preps
Sample Source Gels and Solutions

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.