Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RPM™ Kit

Catalog No. MP112070500 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
120 Preps
300 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP112070500 300 Preps
MP112070400 120 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP112070500 Supplier MP Biomedicals™ Supplier No. 112070500

missing translation for 'validMainframeMsg'

For Nucleic Acid Isolation and Purification, Sample Lysis and Preparation

  • Kit is designed for rapid isolation and purification of double-stranded plasmid DNA from bacterial cultures. Theprocess yields pure plasmid DNA that is suitable for enzymatic manipulations including DNA sequencing, restriction enzymedigestion, in vitro transcription and PCR.
  • For research use only
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For rapid isolation, purification of double-stranded plasmid DNA from bacterial culture
Includes Pre-Lysis, Alkaline Lysis, Neutralizing, Glassmilk spin buffers, wash soln. concentrate
No. of Reactions 300 Preps
Product Type RPM Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.