Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Sheep Serum

Catalog No. ICN642951 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN642951 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN642951 Supplier MP Biomedicals™ Supplier No. 08642951
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile, filtered, Lyophilized Serum

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Species Sheep
Form Lyophilized
Product Type Sheep Serum
Quantity 500 mL
Concentration 7.2g/dL
For Use With (Application) Propagation of long-term cultures of erythrocytic stages of the Theileria uilenbergi intracellular protozoan parasite
Lyophilized Yes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.