Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPIN Module and Recovery Tubes

Catalog No. 112080800 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Pk.
60 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-208-0800 100 Pk.
11-208-0600 60 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-208-0800 Supplier MP Biomedicals™ Supplier No. 112080800
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for optimal performance, providing the purest DNA faster than ever, and with no matrix carry-over. SPIN Module and Recovery Tubes are applied in molecular biology for their ability to separate particles from solutions in a few seconds.

  • Spin Module give faster and cleaner separations than traditional methods such as columns, filters, or simple centrifugation
  • When used with the GENECLEAN™ or MERmaid™ protocols, they combine the speed, efficiency, and selectivity of the GLASSMILK™ binding reaction with an even more rapid execution of the separation, washing, and elution steps
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Tube
Isolation Technology Glass Milk
Sample Type Lysate or other suspension
For Use With (Application) SPIN Module is applied in molecular biology for its ability to separate particles from solutions in a few seconds.
Product Type SPIN Module and Recovery Tubes
Quantity 100 Pk.
Sample Volume 500 to 700 μL liquid per centrifugation, Larger volumes require multiple loadings
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.