Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Stem Cell Factor, Human, Recombinant

Catalog No. ICN15869810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 μ g
2μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15869810 10 μ g
ICN15869802 2μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15869810 Supplier MP Biomedicals™ Supplier No. 0215869810
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stimulates the proliferation of bone marrow cells in 5-fluorouracil treated mice

  • Hematopoietic growth factor, also known as Mast Cell Growth Factor or c-kit ligand
  • Expressed in E. coli
  • ED50 is 20.0 ng/mL

Believed to stimulate the proliferation of myeloid, erythroid and lymphoid progenitors in bone marrow cultures and acts synergistically with colony stimulating factors, resulting in greater size and yields

Order Info

Supplied lyophilized

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Stem Cell Factor
Formulation Sterile filtered then the protein was lyophilized with no additives
For Use With (Application) Believed to stimulate the proliferation of myeloid, erythroid and lymphoid progenitors in bone marrow cultures and acts synergistically with colony stimulating factors, resulting in greater size and yields
Purity or Quality Grade ≥98%
Quantity 10 μ g
Reactivity E. coli
Source Human
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.