Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ T-Butylated Hyrdoquinone

Catalog No. ICN96037480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN96037480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN96037480 Supplier MP Biomedicals™ Supplier No. 0296037480

missing translation for 'validMainframeMsg'

Purity: 99%

  • Molecular Formula: C10H14O2
  • Molecular Weight: 166.22
  • CAS No.: 1948-33-0
  • Synonym: BHQ; 2,5-Di-tert-butylhydroxyquinone; 2-(1,1-Dimethylethyl)-1,4-Benzenediol
  • Physical Description: White to pink crystalline powder
  • Description: CAO-3 Antioxidant is the food grade desgnation for 2,6-di-tert-butyl-para-cresol (BHT Butylated Hydroxytoluene). CAO-3 is a direct replacement for CAO-1 (technical grade) BHT
  • Solubility: Soluble in alcohol, ether and methanol, acetone; slightly soluble in water (0.5%)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Diet Supplement
Particle Shape Powder
Quantity 100 g
Species Swine
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.