Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TallPrep™ Tubes, 4.5 mL, Empty

Catalog No. MP116970000 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116970000 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116970000 Supplier MP Biomedicals™ Supplier No. 116970000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Empty TallPrep™ Tubes for use in Nucleic Acid Isolation and Purification.

  • 4.5 mL TallPrep™ Tubes (Empty)
  • Caps included
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Sample Type All
Includes Caps
Product Type Empty Tube
Quantity 100 Pk.
Volume (Metric) 4.5 mL

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.