Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TCM Defined Serum Replacement

Catalog No. ICN2010026 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN2010026 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN2010026 Supplier MP Biomedicals™ Supplier No. 092010026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fortified, multipurpose serum replacement for the long-term culturing of anchorage dependent and suspension cells from a variety of species.

TCM is a fortified, multipurpose serum replacements for the long-term culturing of anchorage dependent and suspension cells from a variety of species. These serum replacements are effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.

  • Universal serum replacement that supports human and mammalian cells
  • Completely free of virus, mycoplasma and other infectious agents
  • Contains no growth factors, steroid hormones or phenol red
  • Highly purified and heat treated bovine albumin and transferrin are added with final total protein concentration less than 0.1%
  • Packaged as a 50X concentrate; therefore, 10mL will produce 500mL of growth medium

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Product Type FBS Replacement Serum
Quantity 20 mL
pH 6.8 to 7.2
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.