Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Whole Serum, Hamster, Purified, 2mL

Catalog No. ICN55986 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55986 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55986 Supplier MP Biomedicals™ Supplier No. 0855986

missing translation for 'validMainframeMsg'

Whole Hamster Serum with buffer salts

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Species Hamster
Product Type Hamster Serum
Quantity 2 mL
Sterilization Method 0.2 μm filtration
For Use With (Application) Serum Protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques, used as a blocking agent in fluorescence and enzyme immunoassays
Lyophilized Yes
pH 7.3
Protein Content 62.1mg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.