Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Whole Serum, Horse, Purified, 2mL

Catalog No. ICN55987 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55987 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55987 Supplier MP Biomedicals™ Supplier No. 0855987
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Whole Horse Serum with buffer salts

TRUSTED_SUSTAINABILITY

Specifications

Species Horse
Form Lyophilized
Product Type Horse Serum
Quantity 2 mL
Sterilization Method 0.2 μm filtration
Formulation Delipidated, lyophilized from PBS with Azide
For Use With (Application) Immunoelectrophoresis, ouchterlony immunodiffusion, and other immunoprecipitation techniques, blocking agent in fluorescence and enzyme immunoassays
pH 7.3
Protein Content 63.5 mg/mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.