Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Whole Serum, Monkey, Purified, 2mL

Catalog No. icn55988 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55988 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55988 Supplier MP Biomedicals™ Supplier No. 0855988
Only null left
Add to Cart
Add to Cart

Whole Monkey Serum with buffer salts

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Species Monkey
Product Type Monkey Serum
Quantity 2 mL
For Use With (Application) Serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques
Lyophilized Yes
Protein Content 63.9mg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.