Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ X-Gal Powder

Catalog No. MP114063102 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114063102 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114063102 Supplier MP Biomedicals™ Supplier No. 114063102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to detect β-galactosidase activity to differentiate recombinants from nonrecombinants in cloning experiments

Protein trafficking; Cell viability; Cytotoxicity;Gene expression; Transformation; Cloning and expression; Bacteria cell culture; Microbiological analysis

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15° to 30°C
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.