Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Yeast Cell Lysis Preparation Kit

Catalog No. MP112015600 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
100 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP112015600 100 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112015600 Supplier MP Biomedicals™ Supplier No. 112015600

missing translation for 'validMainframeMsg'

Designed for yeast lysis

The kit includes Yeast Suspension Buffer, Yeast Enzyme Enhancer, Spheroplast Indication Solution, Spheroplast Control Solution, Yeast Enzyme Salts, Spheroplast Enzyme Solution and Spheroplast Enzyme Mix
TRUSTED_SUSTAINABILITY

Specifications

No. of Reactions 100 Preps
Product Type Yeast cell lysis preparation kit

Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.