Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YPD Sterile Solution

Catalog No. MP114001034 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP114001034 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114001034 Supplier MP Biomedicals™ Supplier No. 114001034
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rich media for the growth of S. cereviseae in liquid medium

  • Contains dextrose, peptone, and yeast extraction
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose
  • pH 6.5 at 30°C
  • Sterile solutions

Yeast Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Appearance Pale Yellow
Broth Type YPD Broth
Certifications/Compliance Molecular Biology Certified
Concentration 50 g/L
Content And Storage 15° to 30°C
Form Solution
Media Type YPD
Product Type Microbiology Media
Sterility Sterile
Target Organism Class Yeast
Test Type Growth Promotion Testing
Fill Volume 1 L
Formulation Contents/L-20 g Peptone Y, 10 g Yeast Extract-Y, 20 g Dextrose, Biological Buffers
For Use With (Application) Saccharomyces cerevisiae
Packaging Type Bottle
pH 6.5
Quantity 1 L
Source Yeast
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.