Learn More
MSC MP CLEANERDEGREASER
Catalog No. 502818403
Encompass
Apriza II is a concentrated, oxidizing multi-purpose cleaner, degreaser, sanitizer, and deodorizer for hard, non-porous surfaces, designed for light to heavy duty cleaning tasks.
  • Oxidizing multi-purpose cleaner, degreaser, and sanitizer for non-food contact surfaces
  • Deodorizes and eliminates offensive odors
  • Sanitizes hard, non-porous surfaces within 5 minutes
  • Effective for general wipe down cleaning, mopping solutions, and carpet spot removal
  • Leaves a pleasant lemongrass fragrance
  • Concentrated formula allows for varying dilution rates for economical use
  • VOC compliant and contains peroxide for enhanced cleaning
  • Low pH (3.5-4.0) mild acidic formulation prevents alkaline damage
  • EPA registered as a sanitizer
Catalog No. 50-281-8403 Supplier MSC Supplier No. 17643131
Apriza II is a concentrated, oxidizing multi-purpose cleaner, degreaser, sanitizer, and deodorizer for hard, non-porous surfaces, designed for light to heavy duty cleaning tasks.
  • Oxidizing multi-purpose cleaner, degreaser, and sanitizer for non-food contact surfaces
  • Deodorizes and eliminates offensive odors
  • Sanitizes hard, non-porous surfaces within 5 minutes
  • Effective for general wipe down cleaning, mopping solutions, and carpet spot removal
  • Leaves a pleasant lemongrass fragrance
  • Concentrated formula allows for varying dilution rates for economical use
  • VOC compliant and contains peroxide for enhanced cleaning
  • Low pH (3.5-4.0) mild acidic formulation prevents alkaline damage
  • EPA registered as a sanitizer