Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC mp-dLAE-PABC-MMAE | 2625616-44-4 | 98.3% | 1331.59 g·mol⁻1 | C68H102N10O17 | 10 MG
SDP

Catalog No. 5000286163
Encompass_Preferred

mp-dLAE-PABC-MMAE is a drug-linker conjugate designed for use in antibody-drug conjugate (ADC) synthesis. The compound incorporates monomethyl auristatin E as the cytotoxic payload and a para-aminobenzyl carbamate (PABC) cleavable spacer, and is supplied in milligram quantities for research and development.

  • Contains monomethyl auristatin E, a potent tubulin inhibitor.
  • Includes a PABC cleavable linker for enzyme-triggered payload release.
  • Supplied as a solid in multiple milligram sizes suitable for ADC conjugation workflows.
  • Molecular formula C68H102N10O17 and high purity for research applications.
  • Suitable for preclinical ADC synthesis and payload conjugation studies.

Catalog No. 50-002-86163 Supplier Medchemexpress LLC Supplier No. HY15291610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

mp-dLAE-PABC-MMAE is a drug-linker conjugate designed for use in antibody-drug conjugate (ADC) synthesis. The compound incorporates monomethyl auristatin E as the cytotoxic payload and a para-aminobenzyl carbamate (PABC) cleavable spacer, and is supplied in milligram quantities for research and development.

  • Contains monomethyl auristatin E, a potent tubulin inhibitor.
  • Includes a PABC cleavable linker for enzyme-triggered payload release.
  • Supplied as a solid in multiple milligram sizes suitable for ADC conjugation workflows.
  • Molecular formula C68H102N10O17 and high purity for research applications.
  • Suitable for preclinical ADC synthesis and payload conjugation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.