Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP EcoChrome™ Alumina A, Activity I, 63–200μm, MP Biomedicals™

Catalog No. MP0202102 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0202102 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0202102 Supplier MP Biomedicals, Inc Supplier No. 0202102
Add to Cart
Add to Cart

Used in laboratories (e.g. chromatography medium) and for industrial uses as a good all-around adsorbent, desiccating agent and catalyst. MP EcoChrome™ Alumina A, Activity I, 63–200μm, MP Biomedicals can be adjusted to lower activity by t

Alumina A is an oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc. It can be adjusted to lower activity by the addition of polar media, preferably water, but also higher alcohols, e.g. glycol or glycerol can be used. It catalyzes a variety of reactions that are useful industrially.

  • In chemistry laboratories, as a medium for chromatography (available in basic, acidic and neutral formulations), as well as for alcohols, hydrocarbons, steroids, alkaloids and natural pigments
  • As an abrasive, due to its hardness and strength (e.g. as an inexpensive substitute for industrial diamond; used in many types of sandpaper in crystal form)
  • Widely used to remove water from gas streams
  • In the manufacture of dental cements and refractories
  • Neutral alumina, (less active than the basic form) is useful for separation of aldehydes, ketones, quinones, esters, lactones, and glucosides
  • Acidic alumina (pH 4.5 when in water), is the weakest and least often used type
  • Alumina is useful for separation of acid pigments and strong acids
  • It can cause polymerization, condensation and dehydration reactions
  • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification
  • As well, it is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water
Formula Weight 101.96
Recommended Storage Room temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.