Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alumina N, 32 to 63 μm, Active, MP EcoChrom™, MP Biomedicals™

Catalog No. MP0202062 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0202062 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0202062 Supplier MP Biomedicals, Inc Supplier No. 0202062
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as an adsorbent, desiccating agent, and catalyst. EcoChrom™ Alumina N is an amphoteric oxide that is used in a variety of reactions that are useful industrially. It is commonly referred to as alumin.

Alumina N is an oxide of aluminum, occurring in nature as various minerals such as bauxite, corundum, etc. It catalyzes a variety of reactions that are useful industrially.

  • In chemistry laboratories, as a medium for chromatography (available in basic, acidic and neutral formulations), as well as for alcohols, hydrocarbons, steroids, alkaloids and natural pigments.
  • As an abrasive, due to its hardness and strength (e.g. as an inexpensive substitute for industrial diamond; used in many types of sandpaper in crystal form).
  • Widely used to remove water from gas streams.
  • In the manufacture of dental cements and refractories.
  • Neutral alumina, (less active than the basic form) is useful for separation of aldehydes, ketones, quinones, esters, lactones, and glucosides.
  • Acidic alumina (pH 4.5 when in water), is the weakest and least often used type.
  • Alumina is useful for separation of acid pigments and strong acids.
  • It can cause polymerization, condensation and dehydration reactions.
  • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification.
  • It is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 32 to 63 μm
Technique Chromatography
Quantity 500 g
Content And Storage Store at room temperature (15°C to 30°C).
Product Type Insoluble in water
Material Alumina
Physical Form Powder
MDL Number MFCD00003424
CAS 1344-28-1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.