Learn More
EMD MILLIPORE 3P BIOPHARM Gamma Sterilized Millipak Final Fill 20 Filter Unit | 0.22um | 3/4  in.
Supplier:  EMD MILLIPORE 3P BIOPHARM MFGL02SF3
Encompass
Millipak Final Fill capsules are designed for reliable sterile filtration of small volume, high value solutions. In final filling it is critical to maximize product recovery and maintain sterility. The capsule's stacked disc design minimizes hold up volume over standard pleated devices, increasing product recovery. These user-friendly filters contain a multi-purpose port that simplifies venting, integrity testing and sampling and is validated to maintain an aseptic flowpath even after multiple actuations. Millipak Final Fill capsules contain the proven and trusted Durapore membrane in multiple pore sizes offering flexibility for your specific process needs.
Catalog No. NC1806061