Learn More
VAI Sterile MP Hood, Integrated Face Mask
Catalog No. 19546536 Shop All VAI Products
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-546-536 Small/Medium
19-546-537 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-546-536 Supplier VAI Supplier No. 1700PHMS17037
Add to Cart
Add to Cart
Microporous hood with integrated face mask is individually packaged and suitable for ISO 5 cleanroom environments.
  • Heavy weight, non-woven and high quality microporous (MP) coated material
  • Suited for use where maximum protection to product or wearer is required
  • Liquid resistance, especially to chemicals
Specifications
Product Type Hood with Integrated Face Mask
Size Small/Medium
Material Polypropylene
Color White
Closure Type Pull-over
Disposable Single-use
Model 1700P-HM-S-17037
Sterility Sterile
Packaging Type Individually Packaged
Resistance Liquids
Hood Type Integrated Mask
Cleanroom Class Class 100 (ISO 5)