Learn More
Cayman Chemical mP InhIbItr II 1mg
SDP
Catalog No. 502972349
Encompass_Preferred
A reversible, broad-range inhibitor of MMP-1 (IC50 = 24 nM), MMP-3 (IC50 = 18.4 nM), MMP-7 (IC50 = 30 nM), and MMP-9 (IC50 = 2.7 nM)
Catalog No. 50-297-2349 Supplier Cayman Chemical Supplier No. 159581 MG
A reversible, broad-range inhibitor of MMP-1 (IC50 = 24 nM), MMP-3 (IC50 = 18.4 nM), MMP-7 (IC50 = 30 nM), and MMP-9 (IC50 = 2.7 nM)