Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MP Magnetic Rack 8

Catalog No. MP116570426 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Capacity:
8 x 1.5 to 2 mL tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
MP116570426 8 x 1.5 to 2 mL tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116570426 Supplier MP Biomedicals™ Supplier No. 116570426
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This tool is for fast and easy extraction of nucleic acids using a magnetic method. It is commonly used in combination with MagBeads FastDNA™ kits.

  • Magnet made of high-energy neodymium
    • Small, strong and easy to handle
  • For fast and efficient magnetic separations
  • Design allows easy removal of rack from magnet
    • Tubes can be placed on shaker or vortex during extraction process
    • Good visibility of samples
  • Up to 8 tubes of 1.5mL to 2mL with an outer diameter of less than 10.5 mm 

Storage:

  • Magnet:
    • Stored in a dry and cool place
    • Never use > 50 °C
  • Tube rack:
    • Store at room temp
    • Can be placed with tubes at temp as low as -20°C for a limited time

Cleaning:

  • Use with one of the following solutions:
    • 70% (v/v) isopropyl alcohol
    • 1% (v/v) sodium hypochlorite solution (bleach)
    • 0.1 N HCl solution

Notes:

  • Avoid prolonged exposure of rack to water or aqueous solutions
  • Not autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Capacity 8 x 1.5 to 2 mL tubes
Content And Storage Store magnet in dry, cool place, Store rack at room temp
Isolation Technology Magnetic Bead
Final Product Type Magnetic Rack
For Use With (Equipment) For use with the Magbeads FastDNA™ kit, tubes
Product Type Magnetic Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.