Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ MP-MRC-5™ Medium, Serum-free

Catalog No. MP92013426 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP92013426 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP92013426 Supplier MP Biomedicals™ Supplier No. 092013426

missing translation for 'validMainframeMsg'

Very low protein (90ng/mL) medium developed specifically for the culture of MRC-5 cells.

MP-MRC-5 medium was developed specifically for microcarrier culture of MRC-5 cells in production systems. It is completely animal component-free and contains very little protein. MP-MRC-5 achieves growth rates superior to standard serum-supplemented media. Adaptation from serum-supplemented media is not necessary.

Recommended Storage

Store at +4°C. Protect from light. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line MRC-5 cells
Classification Very Low Protein (<90ng/mL)
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile
With Additives Inorganic Salts, Essential and nonessential amino acids, Peptide supplements of plant origin, Vitamins, Growth Factors, Other organic compounds, Trace elements
Without Additives No Glutamine, No Animal Derived Components
For Use With (Application) Intended for use as a production medium to support the growth of MRC-5 cells in agitated (microcarrier) cultures
Quantity 1 L
pH 7
Content And Storage Store at +4°C. Protect from light. Do not freeze.
Description MP-MRC-5 Medium
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.