Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Silitech, 18-32 μm, MP Biomedicals™

Catalog No. MP0202778 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP0202778 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0202778 Supplier MP Biomedicals, Inc Supplier No. 0202778
Add to Cart
Add to Cart

Specifications

Boiling Point 2230°C
Chemical Name or Material MP Silitech
Melting Point 1710°C
CAS 7631-86-9
pH 7.0 ± 0.5
Physical Form Powder
Quantity 25 kg
Molecular Formula SiO2
Synonym Silica; Dioxosilane; Amorphous silica; Crystalline silica; Silicon dioxide; MPLC; Medium Pressure Liquid Chromatography
Molecular Weight (g/mol) 60.08

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.