Learn More
Bio Rad Laboratories MP TGX 10%, 12 W, 10
Catalog No. NC1085844
Encompass
Pkg of 10, 10% precast polyacrylamide gel, 8.6 x 6.7 cm (W x L), for use with Mini-PROTEAN Electrophoresis Cells
Catalog No. NC1085844 Supplier Bio Rad Laboratories Supplier No. 4568035
Pkg of 10, 10% precast polyacrylamide gel, 8.6 x 6.7 cm (W x L), for use with Mini-PROTEAN Electrophoresis Cells