Learn More
Bio Rad Laboratories MP TGX 4-15%, 12 W, 10
Catalog No. NC1085848
Encompass
Pkg of 10, 4-15% precast polyacrylamide gel, 8.6 x 6.7 cm (W x L), for use with Mini-PROTEAN Electrophoresis Cells
Catalog No. NC1085848 Supplier Bio Rad Laboratories Supplier No. 4568085
Pkg of 10, 4-15% precast polyacrylamide gel, 8.6 x 6.7 cm (W x L), for use with Mini-PROTEAN Electrophoresis Cells