Learn More
Bio Rad Laboratories MP TGX STAIN FREE 10%, 10W, 50
Catalog No. NC1085832
Encompass
Pkg of 10, 10% precast polyacrylamide gel, 8.6 x 6.7 cm (W x L), for use with Mini-PROTEAN electrophoresis cells and stain-free enabled imagers
Catalog No. NC1085832 Supplier Bio Rad Laboratories Supplier No. 4568034
Pkg of 10, 10% precast polyacrylamide gel, 8.6 x 6.7 cm (W x L), for use with Mini-PROTEAN electrophoresis cells and stain-free enabled imagers