Learn More
Bio Rad Laboratories 4–15% Mini-PROTEAN TGX Stain-Free Protein Gels, 10 well, 50 uL
Catalog No. NC1085831
Encompass
Pkg of 10, 4–15% precast polyacrylamide gel, 8.6 × 6.7 cm (W × L), for use with Mini-PROTEAN Electrophoresis Cells.
Catalog No. NC1085831 Supplier Bio Rad Laboratories Supplier No. 4568084
Pkg of 10, 4–15% precast polyacrylamide gel, 8.6 × 6.7 cm (W × L), for use with Mini-PROTEAN Electrophoresis Cells.