Learn More
Selleck Chemical LLC MP07-66-E0956-25MG
SDP
Catalog No. 503137243
Encompass_Preferred
MP07-66 a FTY720 analogue is devoid of immunosuppressive effects and shows promising antitumor effects in chronic lymphocytic leukemia by disruption of the SET-PP2A complex leading to PP2A reactivation
Catalog No. 50-313-7243 Supplier Selleck Chemical LLC Supplier No. E095625MG
MP07-66 a FTY720 analogue is devoid of immunosuppressive effects and shows promising antitumor effects in chronic lymphocytic leukemia by disruption of the SET-PP2A complex leading to PP2A reactivation