Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanocs NANOCS Magnetic Particles, Azide Functional, 1 micron, 1 mL

Catalog No. NC3972570
Encompass

These azide functionalized magnetic particles possess multiple azide functional groups on their surface. These azido groups are capable of reacting with linear alkyne, catalyzed by copper ions through click chemistry, or spontaneously with strain-promoted cyclooctyne (e.g., DBCO) without a catalyst. They are suitable for biomolecule separation, imaging, and diagnostic applications.

  • 2 to 5 mg/mL suspension in aqueous solution; other concentrations available upon request.
  • Brownish or dark brown solution.
  • Reactive group: Azide (–N3).
  • Reactive toward: alkyne.
  • Store at 4°C. Do not freeze.

Catalog No. NC3972570 Supplier Nanocs Supplier No. MP1UAZ1 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These azide functionalized magnetic particles possess multiple azide functional groups on their surface. These azido groups are capable of reacting with linear alkyne, catalyzed by copper ions through click chemistry, or spontaneously with strain-promoted cyclooctyne (e.g., DBCO) without a catalyst. They are suitable for biomolecule separation, imaging, and diagnostic applications.

  • 2 to 5 mg/mL suspension in aqueous solution; other concentrations available upon request.
  • Brownish or dark brown solution.
  • Reactive group: Azide (–N3).
  • Reactive toward: alkyne.
  • Store at 4°C. Do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.