Learn More
Perkin Elmer US LLC MP2 integrated 3 channel micro pump for NexION
Supplier:  Perkin Elmer US LLC N8145066
Encompass_Preferred
MP2 integrated three channel precision micro peristaltic pump for NexION Benefits: Pump design extends tubing lifetime Stand alone and integrated versions Chemically resistant Ceramic pins and PTFE-coated metal components are resistant to acids and organic solvents Pump network software control Precision Low pulsation 12 closely-spaced rollers improve signal stability Optimized for all fl ow rates Accurate pumping across a wide range of rates from low 0.20 µL/min to 10 mL/min 1, 2, 3, 4, 6 and 8 channels (stand alone MP2) 3, 4 and 6 channels (integrated MP2)
Catalog No. 50-211-7251
Add to Cart
Request a Quote