Learn More
Perkin Elmer US LLC MP2 integrated 4 channel micro pump for NexION
Supplier:  Perkin Elmer US LLC N8145067
Encompass_Preferred
The MP2 (Micro Peristaltic Pump) provides better stability than standard peristaltic pumps. Roller configuration and precision machining minimizes gaps between rollers, resulting in even pressure points along the pump tubing. Reduced pump pulsation improves signal stability, data quality and detection limits. Benefits: Pump design extends tubing lifetime Stand alone and integrated versions Chemically resistant -Ceramic pins and PTFE-coated metal components are resistant to acids and organic solvents Pump network software control Precision Low pulsation -12 closely-spaced rollers improve signal stability Optimized for all fl ow rates -Accurate pumping across a wide range of rates from low 0. 20 µL/min to 10 mL/min 1, 2, 3, 4, 6 and 8 channels (stand alone MP2) 3, 4 and 6 channels (integrated MP2)
Catalog No. 50-211-7245
Only null left
Add to Cart
Request a Quote