Learn More
Perkin Elmer US LLC MP2 integrated 6 channel micro pump for NexION
Supplier:  Perkin Elmer US LLC N8145068
Encompass_Preferred
MP2 (Micro Peristaltic Pump) for integration into PerkinElmer NexION inductively coupled plasma mass spectrometers (ICP-MS) provide better stability than standard peristaltic pumps.
Catalog No. 50-211-7235
Add to Cart
Request a Quote