Learn More
Perkin Elmer US LLC MP2 Peripump Valve Two Driver Avio 200
Catalog No. 502112340
Encompass_Preferred
Valve two Driver Assembly for Avio 200 MP2 Peripump (includes mounting screws). For upgrading single DXi pump to dual DXi pump for prepFAST.
Catalog No. 50-211-2340 Supplier Perkin Elmer US LLC Supplier No. N0790955
Add to Cart
Add to Cart
Request a Quote
Valve two Driver Assembly for Avio 200 MP2 Peripump (includes mounting screws). For upgrading single DXi pump to dual DXi pump for prepFAST.