Learn More
USA Scientific Inc 1.40 mL alphanumeric U-bottom push cap tube, refill carrier-96
Catalog No. NC9845559
Encompass
1.40 mL alphanumeric polypropylene push cap U-bottom tube (without cap) in refill carrier-96, 10 refills of 96 (960 tubes)
Catalog No. NC9845559 Supplier USA Scientific Inc Supplier No. 17722677
1.40 mL alphanumeric polypropylene push cap U-bottom tube (without cap) in refill carrier-96, 10 refills of 96 (960 tubes)