Learn More
ATCC MP41; Uveal Melanoma; Human (Homo sapiens)
Catalog No. 502382917
Encompass
MP41 is an epithelial-like cell line that was isolated in 2006 from the primary tumor in the uveal of a 49-year-old, female patient with melanoma. This cell line was deposited by F Nemati, Institute Curie.
Catalog No. 50-238-2917 Supplier ATCC Supplier No. CRL3297
MP41 is an epithelial-like cell line that was isolated in 2006 from the primary tumor in the uveal of a 49-year-old, female patient with melanoma. This cell line was deposited by F Nemati, Institute Curie.