Learn More
ATCC MP46; Uveal Melanoma; Human (Homo sapiens)
Supplier:  ATCC CRL3298
Encompass
MP46 is an epithelial-like cell line that was isolated in 2006 from the primary tumor in the uveal of a 69-year-old, male patient with melanoma. This cell line was deposited by F Nemati, Institute Curie.
Catalog No. 50-238-4417