Learn More
Medchemexpress LLC mP6 (TFA) | 2700321-79-3 | 990.15 | 1 MG
SDP
Catalog No. 5000423236
Encompass_Preferred
mP6 (Myr-FEEERA-OH) is a myristoylated peptide that functions as an Integrin β(3) Inhibitor. It inhibits the interaction of Gα13 with integrin β3 without disrupting talin-dependent integrin function. Additionally, mP6 blocks the GTP usage of Rac1, Rap1, and Rab7, which effectively inhibits the infection of CHO-A24 cells.
  • Myristoylated peptide
  • Functions as an Integrin β(3) Inhibitor
  • Inhibits the interaction of Gα13 with integrin β3
  • Does not disrupt talin-dependent integrin function
  • Blocks the GTP usage of Rac1, Rap1, and Rab7
  • Effectively inhibits the infection of CHO-A24 cells
Catalog No. 50-004-23236 Supplier Medchemexpress LLC Supplier No. HYP10038A1MG
mP6 (Myr-FEEERA-OH) is a myristoylated peptide that functions as an Integrin β(3) Inhibitor. It inhibits the interaction of Gα13 with integrin β3 without disrupting talin-dependent integrin function. Additionally, mP6 blocks the GTP usage of Rac1, Rap1, and Rab7, which effectively inhibits the infection of CHO-A24 cells.
  • Myristoylated peptide
  • Functions as an Integrin β(3) Inhibitor
  • Inhibits the interaction of Gα13 with integrin β3
  • Does not disrupt talin-dependent integrin function
  • Blocks the GTP usage of Rac1, Rap1, and Rab7
  • Effectively inhibits the infection of CHO-A24 cells