Learn More
Medchemexpress LLC Mp6 (TFA) | 98.5% | 5 MG
SDP
Catalog No. 5000423674
Encompass_Preferred
mP6 (Myr-FEEERA-OH) is a myristoylated peptide and an integrin β3 inhibitor. It prevents the interaction of Gα13 with integrin β3 while preserving talin-dependent integrin function. This peptide also blocks the GTP usage of Rac1, Rap1, and Rab7, which effectively inhibits the infection of CHO-A24 cells.
  • Myristoylated peptide
  • Inhibits Gα13-integrin β3 interaction
  • Preserves talin-dependent integrin function
  • Blocks GTP usage of Rac1, Rap1, and Rab7
  • Inhibits infection of CHO-A24 cells
  • For research use only
Catalog No. 50-004-23674 Supplier Medchemexpress LLC Supplier No. HYP10038A5MG
mP6 (Myr-FEEERA-OH) is a myristoylated peptide and an integrin β3 inhibitor. It prevents the interaction of Gα13 with integrin β3 while preserving talin-dependent integrin function. This peptide also blocks the GTP usage of Rac1, Rap1, and Rab7, which effectively inhibits the infection of CHO-A24 cells.
  • Myristoylated peptide
  • Inhibits Gα13-integrin β3 interaction
  • Preserves talin-dependent integrin function
  • Blocks GTP usage of Rac1, Rap1, and Rab7
  • Inhibits infection of CHO-A24 cells
  • For research use only