Learn More
AdipoGen MPAC-Br
SDP
Supplier:  AdipoGen CDXB0018M050
Encompass_Preferred
Chemical. CAS 177093-58-2. Formula C20H20BrNO2. MW 386.28. Synthetic. HPLC-precolumn-derivatization reagent for carboxylic acids. Compared to classical BMC it emits at longer wavelength and has stronger fluorescence.
Catalog No. 50-256-1711