Learn More
Description
- mPAGE 4-12% Bis-Tris Precast Gel, 10x8 cm
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Specifications
Specifications
| Buffer | MES or MOPS Running Buffer |
| Concentration | 4 to 12% Bis to Tris |
| Gel Percentage | 4 to 12% |
| Gel Thickness | 1 mm |
| Gel Type | Bis-Tris |
| Sample Loading Volume | 60 μL |
| Separation of | Protein |
| Wells | 12-well |
| Quantity | 10 Pk. |
| Length (Metric) | 10 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.