Learn More
Chemglass Life Sciences MPC Body x MPC Back-to-Back Body Adapter - Polycarbonate MPC-to-MPC quick-connect coupling for tubing assemblies
SDP
Catalog No. NC2094751
Encompass
The MPC Body x MPC Back-to-Back Body Adapter is a compact, non-valved quick-connect coupling designed to join two MPC coupling halves in a back-to-back configuration, providing a low-profile, kink-resistant connection for single-use and bioprocess tubing assemblies.
  • Polycarbonate construction with purple tint and USP Class VI compatibility.
  • Allows coupling halves to rotate when connected to reduce tube kinks.
  • Suitable for bioprocess and single-use assemblies where compact connections are required.
  • Sterilizable by autoclave, EtO, e-beam, or gamma sterilization methods.
  • Packaged in quantities of 25 for assembly and production use.
This item is not returnable. View return policy
Catalog No. NC2094751 Supplier Chemglass Life Sciences Supplier No. CLS91407003
The MPC Body x MPC Back-to-Back Body Adapter is a compact, non-valved quick-connect coupling designed to join two MPC coupling halves in a back-to-back configuration, providing a low-profile, kink-resistant connection for single-use and bioprocess tubing assemblies.
  • Polycarbonate construction with purple tint and USP Class VI compatibility.
  • Allows coupling halves to rotate when connected to reduce tube kinks.
  • Suitable for bioprocess and single-use assemblies where compact connections are required.
  • Sterilizable by autoclave, EtO, e-beam, or gamma sterilization methods.
  • Packaged in quantities of 25 for assembly and production use.