Learn More
LGC GENOMICS LLC (Lucigen) MPC Protein Precipitation Solution, 500 mLA simple solution and method for the removal of proteins from cell lysates prior to nucleic acid purification. To be used with the MasterPure Kits
Catalog No. NC9913537
Encompass
This reagent is designed for use with the MasterPure™ kit protocols only.
This item is not returnable. View return policy
Catalog No. NC9913537 Supplier LGC GENOMICS LLC Supplier No. MMP095H
This reagent is designed for use with the MasterPure™ kit protocols only.