Learn More
COLDER PRODUCTS COMPANY Adapter Body Colder Products MPC body x MPC back-to-back Polysulfone USP Class VI ADCF Amber tint Free floating Pressure range up to 60 psi 4 1 bar Temp range -40 to 149 deg C Gamma autoclave sterilization
Catalog No. 503131419
Encompass_Preferred
Adapter Body Colder Products MPC body x MPC back-to-back Polysulfone USP Class VI ADCF Amber tint Free floating Pressure range up to 60 psi 4 1 bar Temp range -40 to 149 deg C Gamma autoclave sterilization
Catalog No. 50-313-1419 Supplier COLDER PRODUCTS COMPANY Supplier No. MPC17C1739
Adapter Body Colder Products MPC body x MPC back-to-back Polysulfone USP Class VI ADCF Amber tint Free floating Pressure range up to 60 psi 4 1 bar Temp range -40 to 149 deg C Gamma autoclave sterilization