Learn More
COLDER PRODUCTS COMPANY CPC Coupling Cap Amber tint Polysulfone USP Class VI ADCF Pressure Up to 60 psi 4 1 bar Sterilization Method Gamma Autoclave Temperature Max 300 deg F Temperature Min -40 deg F Temperature Max 149 deg C
Catalog No. 503131425
Encompass_Preferred
CPC Coupling Cap Amber tint Polysulfone USP Class VI ADCF Pressure Up to 60 psi 4 1 bar Sterilization Method Gamma Autoclave Temperature Max 300 deg F Temperature Min -40 deg F Temperature Max 149 deg C
Catalog No. 50-313-1425 Supplier COLDER PRODUCTS COMPANY Supplier No. MPC32039
CPC Coupling Cap Amber tint Polysulfone USP Class VI ADCF Pressure Up to 60 psi 4 1 bar Sterilization Method Gamma Autoclave Temperature Max 300 deg F Temperature Min -40 deg F Temperature Max 149 deg C