Learn More
Accuflow Systems CPC Coupling Body, Polycarbonate, 3/8 In Hose Barb, Non-Valved
Catalog No. NC3440535
Encompass_Preferred
The CPC non-valved in-line coupling body provides a reliable, low-resistance connection for fluid-handling systems. It features a 3/8 in hose barb and is molded from USP Class VI, ADCF polycarbonate compatible with common sterilization methods, supplied in production bags for assembly and single-use workflows.
  • Material: Polycarbonate, USP Class VI, ADCF.
  • Connection: 3/8 in hose barb, in-line coupling body.
  • Valve: Non-valved.
  • Color: Purple tint.
  • Sterilization method: Gamma, autoclave.
  • Packaging: 250 per bag.
Catalog No. NC3440535 Supplier Accuflow Systems Supplier No. MPC17006T03
The CPC non-valved in-line coupling body provides a reliable, low-resistance connection for fluid-handling systems. It features a 3/8 in hose barb and is molded from USP Class VI, ADCF polycarbonate compatible with common sterilization methods, supplied in production bags for assembly and single-use workflows.
  • Material: Polycarbonate, USP Class VI, ADCF.
  • Connection: 3/8 in hose barb, in-line coupling body.
  • Valve: Non-valved.
  • Color: Purple tint.
  • Sterilization method: Gamma, autoclave.
  • Packaging: 250 per bag.