Learn More
Accuflow Systems CPC Back-to-Back Body Adapter, Polysulfone, Non-Valved, Pack of 25
Catalog No. NC3440537
Encompass_Preferred
Polysulfone back-to-back body adapters designed for open-format biopharma connections. Non-valved configuration provides a secure, leak-resistant connection between identical quick-disconnect couplings in single-use fluid systems.
  • Polysulfone material, USP Class VI, ADCF.
  • Non-valved back-to-back body adapter configuration.
  • Natural finish with amber tint.
  • Free floating body, suitable for open-format single-use systems.
  • Pack quantity 25 for system integration and inventory management.
Catalog No. NC3440537 Supplier Accuflow Systems Supplier No. MPC17C1739
Polysulfone back-to-back body adapters designed for open-format biopharma connections. Non-valved configuration provides a secure, leak-resistant connection between identical quick-disconnect couplings in single-use fluid systems.
  • Polysulfone material, USP Class VI, ADCF.
  • Non-valved back-to-back body adapter configuration.
  • Natural finish with amber tint.
  • Free floating body, suitable for open-format single-use systems.
  • Pack quantity 25 for system integration and inventory management.